
(+/-)-2,2-Difluoro-1-methylcyclopropylamine hydrochloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:128230-76-2
:
Formula:C₄H₇F₂N·ClH
MDL Number:MFCD07777162
:143.563 g/mol
Melting Pt:143…146 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together