Methyl 5-(difluoromethyl)-2-(dimethylamino)benzoate
Catalog # APOSPC520314-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:229.223 g/mol
Storage Temperature:Ambient
Formula:C₁₁H₁₃F₂NO₂
MDL Number:MFCD30496627