[4-(Difluoromethyl)-2,6-dimethoxy-phenyl] acetate
Catalog # APOSPC520302-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:246.207 g/mol
Storage Temperature:Ambient
CAS Number:2149601-09-0
Formula:C₁₁H₁₂F₂O₄
MDL Number:MFCD30496621