[4-(Difluoromethyl)-2-ethoxy-phenyl]boronic acid
Catalog # APOSPC520299-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:215.99 g/mol
Storage Temperature:Ambient
Formula:C₉H₁₁BF₂O₃
MDL Number:MFCD30496618