Order Entry
Czech Republic
ContactUsLinkComponent
 
Methyl 2-(difluoromethyl)-3,5-dimethoxy-benzoate
Catalog # APOSPC520295-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 2-(difluoromethyl)-3,5-dimethoxy-benzoate
Catalog # APOSPC520295-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :246.207 g/mol
Storage Temperature:Ambient
CAS Number:2149590-59-8
Formula:C₁₁H₁₂F₂O₄
MDL Number:MFCD30496613