1-(Difluoromethyl)-4-(4-methoxyphenoxy)benzene
Catalog # APOSPC520288-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:250.241 g/mol
Storage Temperature:Ambient
CAS Number:2149601-35-2
Formula:C₁₄H₁₂F₂O₂
MDL Number:MFCD29767772