4-Chloro-5-(difluoromethyl)-2-pyrrolidin-1-yl-thiazole
Catalog # APOSPC520286-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:238.685 g/mol
Storage Temperature:Ambient
Formula:C₈H₉ClF₂N₂S
MDL Number:MFCD29767768