4-[5-(Difluoromethyl)thiazol-2-yl]morpholine
Catalog # APOSPC520283-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:220.24 g/mol
Storage Temperature:Ambient
Formula:C₈H₁₀F₂N₂OS
MDL Number:MFCD29767765