4-[4-Chloro-5-(difluoromethyl)thiazol-2-yl]morpholine
Catalog # APOSPC520282-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:254.685 g/mol
Storage Temperature:Ambient
Formula:C₈H₉ClF₂N₂OS
MDL Number:MFCD29767763