Order Entry
Czech Republic
ContactUsLinkComponent
4-(Difluoromethyl)-2-(4-piperidyl)thiazole hydrochloride
4-(Difluoromethyl)-2-(4-piperidyl)thiazole hydrochloride
Catalog # APOSPC520269-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(Difluoromethyl)-2-(4-piperidyl)thiazole hydrochloride
Catalog # APOSPC520269-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :254.728 g/mol
Storage Temperature:Ambient
Formula:C₉H₁₂F₂N₂S·ClH
MDL Number:MFCD29767750