2,5-Dichloro-3-(difluoromethyl)thiophene
Catalog # APOSPC520260-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:203.037 g/mol
Storage Temperature:Ambient
Formula:C₅H₂Cl₂F₂S
MDL Number:MFCD29767740