2-Bromo-3-(difluoromethyl)thiophene
Catalog # APOSPC520259-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:213.043 g/mol
Storage Temperature:Ambient
CAS Number:2092062-85-4
Formula:C₅H₃BrF₂S
MDL Number:MFCD29767739