Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-(Difluoromethyl)thiophene-3-carbonitrile
5-(Difluoromethyl)thiophene-3-carbonitrile
5-(Difluoromethyl)thiophene-3-carbonitrile
5-(Difluoromethyl)thiophene-3-carbonitrile
APOSPC520234-1G
:
Apollo Scientific
:
5-(Difluoromethyl)thiophene-3-carbonitrile
APOSPC520234-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
159.157 g/mol
:
Ambient
:
C₆H₃F₂NS
:
MFCD29767714