4-Chloro-6-(difluoromethyl)thieno[3,2-d]pyrimidine
Catalog # APOSPC520189-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:220.627 g/mol
Storage Temperature:Ambient
CAS Number:2092627-98-8
Formula:C₇H₃ClF₂N₂S
MDL Number:MFCD29767670