Order Entry
Czech Republic
ContactUsLinkComponent
 
2,3-Dibromo-5-(difluoromethyl)-1H-pyrrole
Catalog # APOSPC520173-1G
Supplier:  Apollo Scientific
CAS Number:  
2,3-Dibromo-5-(difluoromethyl)-1H-pyrrole
Catalog # APOSPC520173-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :274.889 g/mol
Storage Temperature:Ambient
Formula:C₅H₃Br₂F₂N
MDL Number:MFCD29767653