Order Entry
Czech Republic
ContactUsLinkComponent
3-(Difluoromethyl)pyrazolo[1,5-a]pyridine
3-(Difluoromethyl)pyrazolo[1,5-a]pyridine
Catalog # APOSPC520163-250MG
Supplier:  Apollo Scientific
CAS Number:  
3-(Difluoromethyl)pyrazolo[1,5-a]pyridine
Catalog # APOSPC520163-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :168.143 g/mol
Storage Temperature:Ambient
Formula:C₈H₆F₂N₂
MDL Number:MFCD29767643