Order Entry
Czech Republic
ContactUsLinkComponent
 
7-(Difluoromethyl)pyrazolo[1,5-a]pyridine
Catalog # APOSPC520161-1G
Supplier:  Apollo Scientific
CAS Number:  
7-(Difluoromethyl)pyrazolo[1,5-a]pyridine
Catalog # APOSPC520161-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :168.143 g/mol
Storage Temperature:Ambient
Formula:C₈H₆F₂N₂
MDL Number:MFCD29767641