Order Entry
Czech Republic
ContactUsLinkComponent
3-(Difluoromethyl)-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)imidazo[1,2-a]pyridine
3-(Difluoromethyl)-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)imidazo[1,2-a]pyridine
Catalog # APOSPC520154-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(Difluoromethyl)-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)imidazo[1,2-a]pyridine
Catalog # APOSPC520154-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :294.105 g/mol
Storage Temperature:Ambient
Formula:C₁₄H₁₇BF₂N₂O₂
MDL Number:MFCD30496626