5-Bromo-3-(difluoromethyl)-1H-pyrrolo[2,3-c]pyridine
Catalog # APOSPC520132-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:247.04 g/mol
Storage Temperature:Ambient
CAS Number:2090237-62-8
Formula:C₈H₅BrF₂N₂
MDL Number:MFCD29767615