4-Chloro-3-(difluoromethyl)-1H-pyrrolo[2,3-b]pyridine
Catalog # APOSPC520124-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:202.589 g/mol
Storage Temperature:Ambient
CAS Number:2092851-08-4
Formula:C₈H₅ClF₂N₂
MDL Number:MFCD29767607