Order Entry
Czech Republic
ContactUsLinkComponent
 
2-tert-Butyl-3-(difluoromethyl)-5,7-difluoro-1H-indole
Catalog # APOSPC520103-1G
Supplier:  Apollo Scientific
CAS Number:  
2-tert-Butyl-3-(difluoromethyl)-5,7-difluoro-1H-indole
Catalog # APOSPC520103-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :259.243 g/mol
Storage Temperature:Ambient
Formula:C₁₃H₁₃F₄N
MDL Number:MFCD29767586