5-(Difluoromethyl)-2,4-dimethoxy-pyrimidine
Catalog # APOSPC520063-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:190.147 g/mol
Storage Temperature:Ambient
CAS Number:2128733-78-6
Formula:C₇H₈F₂N₂O₂
MDL Number:MFCD29767546