Order Entry
Czech Republic
ContactUsLinkComponent
2-(Difluoromethyl)-6-fluoroquinoline
2-(Difluoromethyl)-6-fluoroquinoline
Catalog # APOSPC520025-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(Difluoromethyl)-6-fluoroquinoline
Catalog # APOSPC520025-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :197.157 g/mol
Storage Temperature:Ambient
Formula:C₁₀H₆F₃N
MDL Number:MFCD29767509