Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl 5-amino-4-bromo-1-(4-fluorophenyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-bromo-1-(4-fluorophenyl)pyrazole-3-carboxylate
Catalog # APOSPC520005-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 5-amino-4-bromo-1-(4-fluorophenyl)pyrazole-3-carboxylate
Catalog # APOSPC520005-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :328.137 g/mol
Storage Temperature:Ambient
CAS Number:1427022-75-0
Formula:C₁₂H₁₁BrFN₃O₂
MDL Number:MFCD23135137