2-Chloro-3-iodo-6-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:205240-59-1
:
Formula:C₆H₂ClF₃IN
MDL Number:MFCD15478058
:307.439 g/mol
Melting Pt:35 °C
Storage Temperature:Ambient
Frequently Bought Together