(1S)-1-(2-Bromophenyl)-N-[(4-fluorophenyl)methyl]ethanamine hydrochloride
Catalog # APOSPC512016-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:344.65 g/mol
Storage Temperature:Ambient
Formula:C₁₅H₁₅BrFN·ClH
MDL Number:MFCD31695785