Order Entry
Czech Republic
ContactUsLinkComponent
N-[(1S)-1-(2-Bromophenyl)ethyl]-2,2,2-trifluoro-ethanamine hydrochloride
N-[(1S)-1-(2-Bromophenyl)ethyl]-2,2,2-trifluoro-ethanamine hydrochloride
Catalog # APOSPC512008-1G
Supplier:  Apollo Scientific
CAS Number:  
N-[(1S)-1-(2-Bromophenyl)ethyl]-2,2,2-trifluoro-ethanamine hydrochloride
Catalog # APOSPC512008-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :318.561 g/mol
Storage Temperature:Ambient
Formula:C₁₀H₁₁BrF₃N·ClH
MDL Number:MFCD31695778