2,2,2-Trifluoro-N-[1-(4-fluorophenyl)ethyl]ethanamine hydrochloride
Catalog # APOSPC512001-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:257.656 g/mol
Storage Temperature:Ambient
Formula:C₁₀H₁₁F₄N·ClH
MDL Number:MFCD31695771