Order Entry
Czech Republic
ContactUsLinkComponent
2,2,2-Trifluoro-N-[(1S)-1-(4-fluorophenyl)ethyl]ethanamine hydrochloride
2,2,2-Trifluoro-N-[(1S)-1-(4-fluorophenyl)ethyl]ethanamine hydrochloride
Catalog # APOSPC512000-1G
Supplier:  Apollo Scientific
CAS Number:  
2,2,2-Trifluoro-N-[(1S)-1-(4-fluorophenyl)ethyl]ethanamine hydrochloride
Catalog # APOSPC512000-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :257.656 g/mol
Storage Temperature:Ambient
Formula:C₁₀H₁₁F₄N·ClH
MDL Number:MFCD31695770