Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Bis(trifluoromethyl)phenyl propyl ether
3,5-Bis(trifluoromethyl)phenyl propyl ether
3,5-Bis(trifluoromethyl)phenyl propyl ether
APOSPC51145-1G
:
Apollo Scientific
:
3,5-Bis(trifluoromethyl)phenyl propyl ether
APOSPC51145-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
272.187 g/mol
:
Ambient
:
1420977-70-3
:
C₁₁H₁₀F₆O
:
MFCD28396341