Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-5-fluoro-4-hydroxypyridine
3-Bromo-5-fluoro-4-hydroxypyridine
3-Bromo-5-fluoro-4-hydroxypyridine
3-Bromo-5-fluoro-4-hydroxypyridine
APOSPC51116-1G
:
Apollo Scientific
:
3-Bromo-5-fluoro-4-hydroxypyridine
APOSPC51116-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
191.986 g/mol
:
GHS07
:
Ambient
:
1214342-60-5
:
C₅H₃BrFNO
:
MFCD13185370