Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl 2-(4-Cyanophenyl)-2,2-difluoroacetate
Ethyl 2-(4-Cyanophenyl)-2,2-difluoroacetate
Katalogové číslo APOSPC510317-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 2-(4-Cyanophenyl)-2,2-difluoroacetate
Katalogové číslo APOSPC510317-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :225.191 g/mol
Storage Temperature:Ambient
CAS Number:854778-10-2
Formula:C₁₁H₉F₂NO₂
MDL Number:MFCD25967876