Ethyl 2,2-Difluoro-2-mesitylacetate
Catalog # APOSPC510313-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:242.262 g/mol
Storage Temperature:Ambient
CAS Number:1436389-43-3
Formula:C₁₃H₁₆F₂O₂
MDL Number:MFCD29065900