Ethyl 2-(4-Biphenylyl)-2,2-difluoroacetate
Catalog # APOSPC510311-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:276.278 g/mol
Storage Temperature:Ambient
CAS Number:73789-98-7
Formula:C₁₆H₁₄F₂O₂
MDL Number:MFCD26728234