Ethyl 2-(4-Acetylphenyl)-2,2-difluoroacetate
Catalog # APOSPC510309-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:242.219 g/mol
Storage Temperature:Ambient
CAS Number:1308915-15-2
Formula:C₁₂H₁₂F₂O₃
MDL Number:MFCD25957738