2-Fluoro-4-morpholinoaniline dihydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2006276-93-1
:
Formula:C₁₀H₁₃FN₂O·2ClH
MDL Number:MFCD29049239
:269.143 g/mol
Storage Temperature:Ambient
Frequently Bought Together