3-(3-Fluorophenyl)-2-hydroxypropionic acid
Catalog # APOSPC510274-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:184.164 g/mol
Storage Temperature:Ambient
CAS Number:246137-12-2
Formula:C₉H₉FO₃
MDL Number:MFCD21296531