Order Entry
Czech Republic
ContactUsLinkComponent
2-(4-Fluorophenyl)-2-hydroxypropionic acid
2-(4-Fluorophenyl)-2-hydroxypropionic acid
Catalog # APOSPC510273-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(4-Fluorophenyl)-2-hydroxypropionic acid
Catalog # APOSPC510273-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :184.164 g/mol
Storage Temperature:Ambient
CAS Number:81170-13-0
Formula:C₉H₉FO₃
MDL Number:MFCD11052520