Order Entry
Czech Republic
ContactUsLinkComponent
3-[(4-Fluorophenyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid
3-[(4-Fluorophenyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid
Catalog # APOSPC510271-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[(4-Fluorophenyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid
Catalog # APOSPC510271-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :321.3 g/mol
Storage Temperature:Ambient
CAS Number:2006277-64-9
Formula:C₁₆H₁₆FNO₅
MDL Number:MFCD28977390