Ethyl 3-[(4-Fluorophenyl)(hydroxy)methyl]isoxazole-4-carboxylate
Catalog # APOSPC510269-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:265.237 g/mol
Storage Temperature:Ambient
CAS Number:2006277-28-5
Formula:C₁₃H₁₂FNO₄
MDL Number:MFCD28977392