2-(Hydroxymethyl)-5-(trifluoromethyl)benzimidazole
Catalog # APOSPC510256-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:216.16 g/mol
Storage Temperature:Ambient
CAS Number:6758-34-5
Formula:C₉H₇F₃N₂O
MDL Number:MFCD12755183