2-(Hydroxymethyl)-5-(trifluoromethoxy)benzimidazole
Catalog # APOSPC510253-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:232.159 g/mol
Storage Temperature:Ambient
CAS Number:919054-04-9
Formula:C₉H₇F₃N₂O₂
MDL Number:MFCD28977362