4,6-Difluoro-2-(hydroxymethyl)benzimidazole
Catalog # APOSPC510252-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:184.143 g/mol
Storage Temperature:Ambient
CAS Number:1505508-34-8
Formula:C₈H₆F₂N₂O
MDL Number:MFCD28977363