5-Fluoro-2-(hydroxymethyl)-4-methylbenzimidazole
Catalog # APOSPC510250-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:180.179 g/mol
Storage Temperature:Ambient
CAS Number:1936068-82-4
Formula:C₉H₉FN₂O
MDL Number:MFCD28977366