1-[4-(Trifluoromethyl)phenyl]pyrazole-4-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1260740-53-1
:
Formula:C₁₁H₇F₃N₂O₂
MDL Number:MFCD15527665
:256.181 g/mol
Storage Temperature:Ambient
Frequently Bought Together