Order Entry
Czech Republic
ContactUsLinkComponent
2,2-Difluoro-2-(2-fluorophenyl)ethanol
2,2-Difluoro-2-(2-fluorophenyl)ethanol
Catalog # APOSPC510209-1G
Supplier:  Apollo Scientific
CAS Number:  
2,2-Difluoro-2-(2-fluorophenyl)ethanol
Catalog # APOSPC510209-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :176.136 g/mol
Storage Temperature:Ambient
CAS Number:1380202-32-3
Formula:C₈H₇F₃O
MDL Number:MFCD28038850