Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Registrovat
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(2-Iodoethyl)benzotrifluoride
4-(2-Iodoethyl)benzotrifluoride
4-(2-Iodoethyl)benzotrifluoride
APOSPC510161-1G
:
Apollo Scientific
:
4-(2-Iodoethyl)benzotrifluoride
APOSPC510161-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
300.059 g/mol
:
Ambient
:
178685-14-8
:
C₉H₈F₃I
:
MFCD18207440