3'-(Dimethylamino)-2,2,2-trifluoroacetophenone
Catalog # APOSPC510160-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Typ balení:Bottle
- Bal.:1 g
- Sekvence:3047
Specifications
About this item
:
:217.188 g/mol
Storage Temperature:Ambient
CAS Number:33284-23-0
Formula:C₁₀H₁₀F₃NO
MDL Number:MFCD11556187