Methyl 1-(3-Fluoro-4-methoxyphenyl)-4-oxocyclohexanecarboxylate
Catalog # APOSPC510124-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:280.291 g/mol
Storage Temperature:Ambient
CAS Number:1385694-75-6
Formula:C₁₅H₁₇FO₄
MDL Number:MFCD22423127