1-(3-Fluoro-4-methoxyphenyl)-4-oxocyclohexanecarboxylic acid
Catalog # APOSPC510123-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:266.265 g/mol
Storage Temperature:Ambient
CAS Number:1385694-60-9
Formula:C₁₄H₁₅FO₄
MDL Number:MFCD22423126